SP Bel-Art Dust Pan, Bel-Art Products, a part of SP
# de catalogue 66104-110
Fournisseur: Bel-Art Products, a Part of SP
Spécifications
- Cat. no.:66104-110
- Handle Length:10.2 cm (4")
- Dimensions:20.3W x 16.5D x 5.7H cm (8 x 61/2 x 21/4")
- Supplier No.:368390000
Spécifications
A propos de cet article
This dust pan features a very thin forward edge for collecting fine dust.
- Polypropylene construction
- Durable and inert
- Deep back allows safe transport of captured waste
- Autoclavable
Molded of inert polypropylene, it is an excellent pick-up pan for spilled chemical wastes, both liquid and semi-solid. A curved rise near the front edge keeps contents from spilling out accidentally and a deep back allows safe transport of wastes for proper disposal. The integral handle makes handling and carrying more comfortable. Inner rounded contours make cleaning easy. Steam autoclavable at 121°C (250°F).
Customers Who Bought This Also Bought