OHAUS® Laboratory Vortex Mixers
Fournisseur: OHAUS CORP.
New Product
Certificats
A propos de cet article
Offering gentle to high-speed reliable mixing.
- Touch mode operation activates mixing when depressing the cup head or continuous mode for use with accessory attachments
- Robust digital and non-digital vortex mixers offer effective speed control from low RPM start-up for gentle shaking to high-speed mixing for vigorous vortexing of samples
- Sturdy design stabilizes mixer at all speeds
- The fixed speed vortex mixer is an economical choice for vigorous mixing of samples at high speed. To ease repetitive sample processing, this unit provides simple, effective full-speed mixing
OHAUS vortex mixer models allow gentle to high-speed reliable mixing. The fixed speed mixer is an economical choice for vigorous mixing of samples at high speed. The analog model offers variable speed control, and the digital model allows input of the exact speed and time to achieve repeatable results every time. The pulsing model features a unique pulse action that reduces heat generation, while providing more effective mixing and cell disruption.
Certifications: Product Safety: CAN/CSA C22.2 61010-1, CAN/CSA C22.2 61010-2-051, UL 61010-1
Electromagnetic Compatibility: ICES-003 Class A, FCC Part 15 Class A
Compliance Marks: TÜV SÜD
Electromagnetic Compatibility: ICES-003 Class A, FCC Part 15 Class A
Compliance Marks: TÜV SÜD
Caution: https://us.ohaus.com/en-us/legal/caprop65
Customers Who Bought This Also Bought