UltraSOLV® ScrubTIPS® Cleaning Swabs with Grit Tips, Foamtec International
# de catalogue 10809-942
Fournisseur: Foamtec International
Spécifications
- Length:15.2 cm (6")
- Description:Silicon carbide grit tip, 600-Grit
- Cat. no.:10809-942
- Tip Style:Rigid
- Supplier No.:HT170060-5
Spécifications
A propos de cet article
These unique cleaning tools are ideal for polishing stains, scratches, and process residue from hard-to-clean surfaces such as electro-static chucks
Swabs are available with a dual tip or rigid tip and washed diamond or silicon carbide abrasives.
Ideal for removing flashing from sensitive medical devices and optics and maintaining high-precision injection molds.
Customers Who Bought This Also Bought