Spécifications
- Description:VersaHOCl™ disinfectant and cleaner
- Packaging:Double bagged
- Volume:1 gal./128 oz.
- Packing Type:Open top bottle
Spécifications
A propos de cet article
VersaHOCl™ is an EPA registered, RTU (ready to use) one-step cleaner, disinfectant, and deodorizer. Made in the USA with no preservatives or fragrances, Berkshire’s VersaHOCl™ is a powerful and safer disinfectant alternative for cleaning and disinfecting needs.
- Kills common microbiological contaminations on surfaces
- Can be used in a variety of applications
- Ideal for large surface cleaning
- No strong odor provides good user acceptability
- No special disposal is required
- Facilitates transfer disinfection into cleanroom
- No mixing required
Non-toxic with low odor and low residue, VersaHOCl™ establishes the safest standards in disinfectant technology. With an active ingredient of 0.065% hypochlorous acid, it’s non-corrosive and effective on hard, non-porous surfaces. This versatile disinfectant is ideal for use in controlled environments such as labs, cleanrooms, compounding pharmacies, and manufacturing facilities. It can also be used in areas such as hospitals, clinics, healthcare facilities, schools, and government and office buildings.