Spécifications
- Description:Compresstome legacy model blade holder
Spécifications
A propos de cet article
Replacement blade holder for the Compresstome vibrating microtome.
- Compatible with models VF-200, VF-200-0Z, VF-300, and VF-300-0Z
A replacement blade holder for legacy model Comprestomes. This device is compatible with Compresstome models VF-200, VF-200-0Z, VF-300, and VF-300-0Z. Includes one blade holder, one small Allen wrench, and one small screw.