Spécifications
- Description:Laboratory hot water bath
- Capacity:16 l
Spécifications
A propos de cet article
Laboratory hot water bath with a 16 liters capacity.
- Use to incubate sections from slicing on a rotary microtome
- Operating temperature up to 100 °C
- Digital temperature display and easy controls
Applications include incubation of tissue slices for electrophysiology and precision-cut tissue slices, and paraffin-embedded wax slices from rotary microtome sectioning.