ContactUsLinkComponent
Laboratory Hot Water Bath
Laboratory Hot Water Bath
# de catalogue 77435-286
New Product
Laboratory Hot Water Bath
# de catalogue 77435-286
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Laboratory hot water bath
  • Capacity:
    16 l
Spécifications
A propos de cet article
Laboratory hot water bath with a 16 liters capacity.
  • Use to incubate sections from slicing on a rotary microtome
  • Operating temperature up to 100 °C
  • Digital temperature display and easy controls
Applications include incubation of tissue slices for electrophysiology and precision-cut tissue slices, and paraffin-embedded wax slices from rotary microtome sectioning.
Customers Who Bought This Also Bought