Compresstome® Magnetic Blade Holder for VF-210-0Z and VF-510-0Z
# de catalogue 77435-420
Fournisseur: PRECISIONARY INSTRUMENTS LLC
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Description:Compresstome VF-210-0Z and VF-510-0Z magnetic blade holder
Spécifications
A propos de cet article
This is a magnetic blade holder for easy stainless steel blade installation. This can also be used for ceramic blades.
- Premium magnetic blade holder for the compresstome vibrating microtome
- Each blade holder has built-in magnets to securely hold stainless steel blades without glue
- Ceramic and tungsten blades can also be used with glue
Packaging: Includes one magnetic blade holder, one allen wrench, and one small screw.
Customers Who Bought This Also Bought