Compresstome® Double Wall Buffer Tank
# de catalogue 77435-434
Fournisseur: PRECISIONARY INSTRUMENTS LLC
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Description:Double wall buffer tank
Spécifications
A propos de cet article
Double-walled, polycarbonate buffer tank.
- Polycarbonate buffer tank
- Rapidly chill and maintain cool temperatures for optimal slicing
- Full one year warranty
This buffer tank is used to add ice around the buffer solution to keep it colder longer for live tissue slicng.
Packaging: Includes double-wall buffer tank, one set of screws, one gasket, and one small allen wrench.
Customers Who Bought This Also Bought