Verispec® Certified Reference Material/Standard, KCl Conductivity Standards at 25°C, RICCA Chemical Company
Supplier: Ricca Chemical
New Product
Certificates
About this item
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility
Caution: Store at room temperature. The conductivity of these certified reference materials are strongly dependent on temperature. All calibrations with these certified reference materials should be conducted at 25°C.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.