Quebec® Dark-Field Colony Counter, Reichert
Fournisseur: Reichert Technologies
Certificats
A propos de cet article
The Quebec® Dark-Field Colony Counter is designed for use in counting bacteria colonies that are presented against a dark background.
Illumination is provided by 40W tungsten bulbs. The 113mm lens offers 1.5x magnification and is positioned to eliminate parallax errors. Focus lens by sliding rod; swing away when not in use. Second optional lens may be attached to increase magnification to 3x. Sheet metal case is finished in contrasting gray-beige acrylic enamel and black suede.
Certifications: CSA certified. UL listed.
Ordering information: Unit is complete with Wolffhuegel guide plate and three-wire cord.
Spécifications
- Catalog No:
- 23610-158
- 23610-157
- Dimensions:
- 26.7L x 27.9D x 25.4H cm (101/2 x 11 x 10")
- 26.7L x 27.9D x 25.4H cm (101/2 x 11 x 10")