Multiplate Developing Chamber
Catalog # 21432-740
Supplier: General Glass Blowing
Specifications
- Description:Chamber
- Cat. no.:21432-740
- W×D×H:27×7.5×26 cm (10.6×3×10.2")
- Supplier No.:80-10
Specifications
About this item
Developing chamber is extremely useful for TLC
Consists of a sturdy, molded clear glass block with flat surface walls and ground lip of uniform flatness. Raised ridge midway on inside ends and bottom of chamber. Holds up to six 20x20cm plates.
Weight: 6.8 kg (15 lbs.)
Ordering information: Includes jar, anodized aluminum rack, and nonslip lid with lifting knob.
Customers Who Bought This Also Bought