Synthware Vacuum Manifold with High Vacuum Valves, Kemtech America
Fournisseur: Kemtech America
CE Compliant
Certificats
A propos de cet article
Vacuum manifolds are available with a serrated hose connection (10 mm ext. Ø) on the right, left, or both right and left hand sides.
- Hi vacuum valves
- Heavy wall borosilicate construction
- 3, 4 or 5 port options available
The main tube has a 22 mm ext. Ø, and the distance between valves is 100 mm center to center. All manifolds feature a stopcock bore of 0 to 4 mm.
Customers Who Bought This Also Bought