Synthware High Vacuum Manifold with Solid Glass Stopcock, Kemtech America
Fournisseur: Kemtech America
CE Compliant
Certificats
A propos de cet article
Manifolds are equipped with 4 mm double oblique, high vacuum solid glass stopcocks which are precision ground with corresponding numbers on barrel and plug.
- Hollow glass stopcocks (held in by vacuum)
- Heavy wall borosilicate construction
- 3, 4 or 5 port options available
Solid glass stopcocks have constant pressure retention devices. Manifold beams have an ext. Ć of 22 mm and are available in a variety of opening configurations. Distance between stopcocks is 100 mm center to center.
Customers Who Bought This Also Bought