Synthware Distillation Adapters with Threaded Thermometer Port, Kemtech America
Fournisseur: Kemtech America
Certificats
A propos de cet article
Adapter accommodates thermometer in threaded port.
- Nylon cap and Viton® O-ring included
- Thermometer not included
Supplied complete with nylon cap and Viton® O-ring to accommodate 6.0 to 7.5 mm ext. Ø plain thermometer.
Ordering information: Thermometer is not supplied.
Customers Who Bought This Also Bought