Human Recombinant TNFRSF17 Protein (from HEK293)
# de catalogue 77474-004
Fournisseur: Creative Biomart
New Product
Numéro CAS:
Spécifications
- Endotoxin-free:No
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Conjugation:Unconjugated
- Size:50 µg
- Tag sequence:C-Fc, N-His
- Storage conditions:Store it under sterile conditions at −20 to −70 °C. It is recommended that the protein be aliquoted for optimal storage. Avoid repeated freeze-thaw cycles.
- Endotoxin content:<1E U/μg by LAL method
- Biological activity:Measured by its binding ability in a functional ELISA. Immobilised recombinant human BAFF at 1 μg/ml (100 μl/well) can bind human TNFRSF17. The EC50 of human TNFRSF17 is 0.0059 μg/ml.
- Reconstitution instructions:It is recommended that sterile water be added to the vial to prepare a stock solution of 0.2 μg/μl. Centrifuge the vial at 4 °C before opening to recover the entire contents.
- Protein/peptide name:TNFRSF17
- Purity:≥95% as determined by reducing SDS-PAGE.
- Molecular weight:The recombinant human TNFRSF17/Fc is a disulfide-linked homodimer. The reduced monomer comprises 302 amino acids after removal of the signal peptide and has a predicted molecular mass of 34 kDa.
Spécifications
A propos de cet article
Recombinant Human TNFRSF17(Met 1- Ala 54)(NP_001183.2) was expressed in HEK293, fused with a polyhistidine-tagged Fc region of human IgG1 at the C-terminus and a signal peptide at the N-terminus.
Customers Who Bought This Also Bought