A propos de cet article
These syringes comes with the three types of seals.
- Front fitting: ¹/₄"-28 UNF
- Glass barrel: Borosilicate 3.3
- Stroke: 60 mm
PTFE-seal: Chemically resistant; heavy duty syringes.
PTFE-seal with silicone: Specially designed for handling of strong buffers and other aqueous solutions. The modified PTFE-seals guaranteed, extended lifetime under heavy duty conditions.
HDPE-seal: Specially designed for handling of strong buffers and other aqueous solutions. The special PE-seals run smoother than conventional seals. Good resistance to buffers and aqueous solutions, long lifetime.