Syringes for Autosampler Dosage, N
Fournisseur: INNOVATIVE LABOR SYSTEME GMBH
New Product
Certificats
A propos de cet article
The N series syringes for Sample dosage for Gas Chromatography (GC), HPLC Valve, autosamplers, Thin Layer Chromatography (TLC).
- Fixed needles
- Stainless steel plunger tip
- Glass Barrel: Borosilicate 3.3
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.