UltraSOLV® 1700 Series Swabs, Large Bulb Head, Foamtec
# de catalogue 10811-144
Fournisseur: Foamtec International
Spécifications
- Cat. no.:10811-144
- Handle Length:152.4 mm (6.000")
- Head Width:10 mm (0.400")
- Head Length:22.2 mm (0.875")
- Handle Width:3.2 mm (0.125")
- Supplier No.:HT1700-500
Spécifications
A propos de cet article
This high-absorbancy, low-extractable foam head swab is ideal for semiconductor and electric assembly applications
Swab cleans, absorbs and removes contaminants from the surfaces of critical electronic devices. Polypropylene handle.
Ordering information: Swabs are packaged and sealed in five polypropylene mesh bags of 100 swabs each, then placed in a clear, cleanroom bag.
Customers Who Bought This Also Bought