A propos de cet article
Series H-KP automatic liquid handling syringe with a glass barrel made from borosilicate 3.3 designed for volumes of 25 µl to 50 ml. 60 mm stroke and the syringe has a PTFE (polytetrafluoroethylene) plunger tip. A high chemical resistance makes this syringe a perfect heavy-duty syringes.
The syringe is manufactured from high-quality DURAN 3.3 borosilicate glass, that is heat and shock resistant, which ultimately protects against breakage from shock and sudden or extreme fluctuations in temperature. This type of ultra-low expansion glass endow the syringe with maximum chemical resistance, outstanding degree of calibration and superior precision.
The ILS H-KP automatic liquid handling syringes can also be used for many liquid handling tasks, such as injecting or dosing, infusing, dispensing reagents, calibration, aspirating, pipetting and sampling.
The plungers in the glass syringe move more smoothly, which leverages to an especially high level of precision. Because of their high precision, great tightness and chemical resistance, ILS micro syringes are chosen whenever heavy-duty syringes are necessary, most commonly used as a high-quality replacement for laboratory and clinical instruments.
Applications: Diagnostics, next generation sequencing, drug discovery, genomics analysis, cancer research, flow cytometry, autosampler for HPLC, gas chromatography, molecular analyzer, lab automation.