Insect Pooter, Safely Collect and Observe Bugs
Supplier: Wards
New Product
Certificates
About this item
The Insect Pooter is an essential entomology apparatus for hands-free collection of insects and other small organisms. It consists of two flexible tubes attached to a clear plastic chamber.
- Ingenious design uses suction to collect and trap insects and small organisms
- Collection method won't pinch or crush specimens
- Mesh covering on aspirator tube prevents specimens from being inhaled
- Observe specimen structure and behavior through the clear viewing chamber
The user positions one tube over the insect and draws it into the viewing chamber by sucking on the other tube. A fine mesh screen blocks the hole leading up into the tube in the user's mouth to prevent the specimen from being accidentally inhaled. Pooters offer an alternative to mechanical collection methods like hand, nets, and tweezers, which can be harmful to the specimen as well as the collector.
Customers Who Bought This Also Bought