Nalgene® Clean Sheets™ Bench and Shelf Liners, Thermo Scientific
Fournisseur: Thermo Fisher Scientific
Certificats
A propos de cet article
Made from white, cross-linked, closed-cell polyethylene foam, liners are 3.1mm (1/8") thick with firm, smooth, matte-white resilient surface.
Liners cushion sensitive instruments and reduce glass breakage
Cut to any size. Liners take staples, tacks, or tape. Supplied in 15.2m (50') rolls.
Use anywhere protection from chemicals, breakage, vibration, heat, or dirt is needed.
Certifications: Liners meet OSHA Standard 29CFR part 1910.1030 for use as protection against bloodborne pathogens.
Customers Who Bought This Also Bought