DURA-STAT® Soft Work Surfaces, Duracote
Supplier: Duracote
Certificates
About this item
These soft, static-dissipative work surfaces have a resistance-to-ground measure of 107 to 108 ohms at 10V, 12% relative humidity
In accordance with the ESD Association, they are tested to EOS/ESD S4.1 to meet industry guidelines for proper electrostatic discharge. Work surfaces are engineered to meet the requirements of MIL-W-87893. The unique construction provides the same resistance-to-ground measurement from any point on the material. Lightweight and flexible, these multi-layer vinyl surfaces are available in light blue or beige. The top surface is semi-conductive, non-tacky, and dirt- and stain-resistant.
Ordering information: Mats are packaged individually. For white mats, contact your Avantor representative.
Customers Who Bought This Also Bought