A propos de cet article
Series XLP Automatic Liquid Handling syringes with a glass barrel made from Borosilicate glass 3.3 designed for volumes of 25 µl to 50 ml.
- Luer or ¼-28 threaded end caps
- Broad variety of syringe interfaces
- Seals available in PTFE, UHMWPE, or custom UHMWPE blends
- Customized designs and materials to suit application requirements
- Stroke 60 mm
A high chemical resistance makes this syringe a perfect heavy-duty syringes. The syringe is heat and shock resistant, which ultimately protects against breakage from shock and sudden or extreme fluctuations in temperature. This type of ultra-low expansion glass endows the syringe with maximum chemical resistance, outstanding degree of calibration and superior precision.
The ILS XLP Syringe Series can also be used for many liquid handling tasks, such as injecting or dosing, infusing, dispensing reagents, calibration, aspirating, pipetting and sampling. ILS syringes are used in applications such as Diagnostics, next generation sequencing, drug discovery, genomics analysis, cancer research, flow cytometry, HPLC, gas chromatography, molecular analyzer and lab automation.
The plungers in the glass syringe move more smoothly, which leverages to an especially high level of precision. Because of their high precision, great tightness and chemical resistance, ILS microsyringes are chosen whenever heavy-duty syringes are necessary; most commonly used as a high-quality replacement in laboratory and clinical instruments.
PTFE seals are chemically resistant and heavy duty syringes. PE seals have long lifetime and are good resistance to strong buffers and other aqueous solution.