NextGen™ V Vial® Crimp-Top Sample Vials, Wheaton, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
A propos de cet article
Sturdy crimp-top vials have a conical interior bottom for maximum retrieval of content.
Vials can be used for small-scale reactions, centrifugation, storage, and shipping. They are ideal for use in the biochemical, pharmaceutical, and radiochemical industries. Fire-glazed bottom protects against glass particulates. Low-particulate packaging protects against contamination during transportation and storage. Clear vials are made from Wheaton 33® low extractable borosilicate glass to safeguard against a change in product pH caused by alkali, which can be extracted by ordinary glass.
Certifications: Vials conform to ASTM Type I, Class A, and USP Type I requirements.
Customers Who Bought This Also Bought