Dry Ice Cold Trap, Welch®, Gardner Denver
# de catalogue 55007-176
Fournisseur: Welch Vacuum
Product available on GSA Advantage®
Spécifications
- Description:Trap, for 1399, 1400, 1405, 8804, 8890, 8905, 8907, 8912, 8917, 8920, 8925 model pumps
- Cat. no.:55007-176
- Supplier No.:1420H-14
Spécifications
A propos de cet article
Dry ice cold trap features a removable 2.8L (3qt.) center well for dry ice/alcohol slurry (do not use acetone) and a visible trapping surface
Trap can maintain cold temperature up to twelve hours depending on vapor load.
ISO size: 12.7 mm (1/2") I.D. Hose
Customers Who Bought This Also Bought