Syringes for Automatic Liquid Handling, XP-XL
Fournisseur: INNOVATIVE LABOR SYSTEME GMBH
New Product
Certificats
A propos de cet article
The XP-XL syringes for automatic liquid handling.
- Front fitting: ¹/₄" to 28 UNF
- Stroke: 30 mm
- Glass barrel: Borosilicate 3.3
PTFE-seal: Chemically resistant; heavy duty syringes.
HDPE-seal: Long lifetime; good resistance to strong buffers and other aqueous solutions.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.