Double-Tube Manifold with Threaded Stopcocks, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
Greaseless, vacuum/gas manifold features a 0 - 4 mm high vacuum
This manifold has Easy-Action, three-way PTFE stopcocks for takeoffs. The bottom and top manifold tubes each feature a #7 Ace-Thred connection for easy cleaning. The ends are supplied with nylon stopper/plugs with FETFE O-Rings (89052-828). The port connections are 8 mm (1/32") O.D. The vacuum and gas connections are 12.7 mm (1/2") O.D. The distance between the takeoffs is 100 mm (315/16"). Fabricated from borosilicate glass.
Ordering information: Available in three or five port combinations.
Customers Who Bought This Also Bought