Synthware Double Manifold with Hollow Glass Stopcocks and Spherical Joints, Kemtech America
Fournisseur: Kemtech America
Certificats
A propos de cet article
Manifolds feature 35/20 ball joints and 4 mm double oblique high vacuum hollow glass stopcocks.
- Heavy wall borosilicate construction
- 4 or 5 port options available
- Spherical joint side connections
- Hollow glass stopcocks (held in by Vacuum)
The stopcocks are precision ground with corresponding numbers on barrel and plugs. Lower vacuum chambers on stopcocks are evacuated with a separate hole in the plug in order to secure the plug in the barrel.
Customers Who Bought This Also Bought