Double-Bank Vacuum/Inert Manifold, Chemglass
Fournisseur: Chemglass
Certificats
A propos de cet article
Double-bank manifold for use with vacuum and inert gas
Valve size is 0 - 4 mm. Both banks have an O.D. of 11 mm. Ports are 100 mm center to center and have 8 mm serrated hose connections. Valves are sealed at a 45° angle for easier manipulation.
Customers Who Bought This Also Bought