KIMBLE® AIRLESS-WARE® Manifold, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
Certificates
About this item
An all glass manifold used to connect inert gas and vacuum sources to sidearm stopcocks (three-place or five-place manifolds)
Each stopcock has a metering groove cut into the plug to provide fine control of vacuum sources and inert gases. Distance between stopcocks is 65 mm (29/16") for the three-place size and 100 mm (315/16") for the five-place size.
Delivery: Supplied with stopcock pressure/vacuum retainers (fits plug sizes: 12/30, 12.60/40, 14.5/50 mm).
Customers Who Bought This Also Bought