Synthware Photochemical Reaction Vessel with Internal Thread, Kemtech America
# de catalogue 60003-762
Fournisseur: Kemtech America
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Spécifications
- Capacity:500 ml
- Supplier No.:V150500
Spécifications
A propos de cet article
Reaction vessel is constructed of borosilicate glass to accommodate an immersion well.
- Heavy wall Borosilicate construction
- Ground glass joints on side arms
- Internally threaded - #50
A #50 top internal thread accepts an RC50 bushing and Viton® O-ring to form a compression seal with the immersion well. The vessel featuers one [ST] 14/20 joint for a sparger tube, one [ST] 24/40 vertical joint for a condenser, and one #7 internal thread sidearm for a thermometer. Supplied complete with quartz immersion well, caps, and O-rings. Accessory items are sold separately.
Customers Who Bought This Also Bought