Inoculating Loops, Platinum, McCormick Scientific
# de catalogue 50807-041
Fournisseur: Leica Biosystems
Spécifications
- Length:45 mm
- Cat. no.:50807-041
- Inner Diameter:5 mm
- Gauge:24
- Supplier No.:39101054
Spécifications
A propos de cet article
Made of platinum wire with 15% iridium.
- Fused loops design
- Platinum and iridium construction
- Inner diameter: 2-5 mm
Loops are 45 mm in length.
Customers Who Bought This Also Bought