Single-Speed Blenders, 1 L, Waring
Fournisseur: Waring Products
Certificats
A propos de cet article
For rapid pulping or mixing
Removable containers of heat-resistant glass or stainless steel have a 1 L working capacity. The 0.5 hp motor revolves blades at 22000 rpm. Enameled metal base.
Widely used for emulsifying and for blending cosmetics, vitamins, paint and varnish, paper, biologicals, and oils.
Certifications: The 120V models are UL listed.
Ordering information: Supplied complete with two-piece lid.
Customers Who Bought This Also Bought