Nunc® Inoculating Loops and Needles, Sterile, Thermo Scientific
Fournisseur: Thermo Fisher Scientific
CE Compliant
Certificats
A propos de cet article
These polystyrene loops and needles are manufactured to very close tolerance levels and surface treated for enhanced droplet adhesion
They provide problem-free streaking with smooth loop surfaces. Ideal for semi-quantitative inoculating procedures. Disposable, nontoxic, and color-coded for product identification. Sterilized by irradiation.
Ordering information: Supplied in easy peel-open bags of 12 or 50.
Customers Who Bought This Also Bought