VWR® Large Pipet Drawer Organizer
# de catalogue 82024-548
Fournisseur: VWR International
Spécifications
- Description:Drawer organizer for large pipets
- Cat. no.:82024-548
- Exterior Dimensions:460×305×65 mm (18×12×2.5")
Spécifications
A propos de cet article
This rugged polystyrene drawer organizer is ideal for storing a variety of glass or plastic pipets and supplies
- Rugged polystyrene construction
- Convenient compact size
- Multiple compartments of varying size
The convenient size fits into standard laboratory drawers or on benches, tabletops, or shelves. Each tray features three compartments of varying size.
Items are kept organized and easily accessible.
First compartment: 43 x 11 cm (17 x 45/16")
Second and third compartment: 43 x 8 cm (17 x 31/8")
Customers Who Bought This Also Bought