Syringes for Automatic Liquid Handling, XP-PD
Fournisseur: INNOVATIVE LABOR SYSTEME GMBH
New Product
Certificats
A propos de cet article
XP-PD syringes made up of Borosilicate 3.3.
- 3 cm stroke
Volume: 12.5 μl to 12.5 ml
Front fitting: ¹/₄" to 28 UNF
Plunger top: 6 to 32 UNC
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.