A propos de cet article
XP series syringes are designed to work with most industry standard syringe pump designs.
- Designed for volumes of 25 µl to 12.5 ml
- Luer or ¹/₄ to 28 threaded end caps
- Glass barrel: Borosilicate 3.3
- Broad variety of syringe interfaces
- Seals available in PTFE, UHMWPE, or custom UHMWPE blends
- Customized designs and materials to suit application requirements
- 3 cm stroke
- A high chemical resistance makes this syringe a perfect heavy-duty syringes
PTFE Seal:
Chemically resistant
Heavy duty syringes
PTFE Black Seals
Specially designed for handling of strong buffers and other aqueous solutions
The modified PFTE-seals guaranteed, extended lifetime under heavy duty conditions
PE Seal
Long lifetime
Good resistance to strong buffers and other aqueous solution
The syringe is manufactured from high-quality borosilicate glass, that is heat and shock resistant, which ultimately protects against breakage from shock and sudden or extreme fluctuations in temperature. This type of ultra-low expansion glass endow the syringe with maximum chemical resistance, outstanding degree of calibration and superior precision.
The ILS XP Syringe Series can also be used for many liquid handling tasks, such as injecting or dosing, however, it can also be applied to remove or inject gas. The most common applications include infusing, dispensing reagents, calibration, aspirating, pipetting and sampling.
The plungers in the glass syringe move more smoothly, which leverages to an especially high level of precision. Because of their high precision, great tightness and chemical resistance, ILS micro syringes are chosen whenever heavy-duty syringes are necessary; most commonly used as a high-quality replacement for laboratory and clinical instruments.
Applications: Syringe pumps for liquid handling applications/pipetting e.g. Tecan Cavro syringe pumps.