System 250 Stereo Zoom Microscopess, LX Microscopes by Unitron
# de catalogue 14218-514
Fournisseur: LX Microscopes by UNITRON
Spécifications
- Description:System 250 Stereo Zoom Microscope
- Cat. no.:14218-514
- Supplier No.:18714
Spécifications
A propos de cet article
The System 250 comes standard with a binocular stereo zoom microscope and two 10x eyepieces
Additional standard components include boom stand with base and two eyepiece shields that enables easy attachment of either a fluorescent ring light or fiber optic ring light.
Ordering information: Additional eyepieces, eyepiece shields, and lenses sold separately.
Customers Who Bought This Also Bought