VWR® Talon® Column Clamps
Supplier: VWR International
Certificates
About this item
Sturdy, easily adjusted, multi-purpose clamps are ideal for holding large cylindrical glassware and similar objects.
- Multi-purpose design
- Sturdy
- Easily adjusted
- Corrosion-resistant stainless steel construction
A large, flat thumbscrew/worm-drive permits maximum tightening or removing in minimum time. Clamps are constructed of corrosion-resistant stainless steel. With 211 mm (85/16") extension arm.
Ordering information: An additional holder must be purchased in order to attach clamps to frames or other apparatus.
Frequently Bought Together