VWR® Talon® Thermometer Swivel Clamp
Catalogue # 21573-060
Fournisseur: VWR International
Product available on GSA Advantage®
Spécifications
- Description:Thermometer Swivel Clamp
- Cat. no.:21573-060
Spécifications
À propos de cet article
Holds 6–8 mm (1/4– 5/16") diameter glass tubes and thermometers 114 mm (41/2") from support rod.
- Safety-adjust spring plate jaws
- Q-clamp design
- Adjustable to any angle with locking wingnut
- Lightweight
- Resistant to corrosion and rust
- Nickel-plated zinc construction
Features safety-adjust spring plate jaws and Q-clamp design. Adjusts to any angle with locking wing nut. Lightweight, rust- and corrosion-resistant. Made of nickel-plated zinc. Built-in holder grips rods to 19 mm (3/4").