VWR® Talon® All-Position Clamp Holder
# de catalogue 21572-658
Fournisseur: VWR International
Product available on GSA Advantage®
Spécifications
- Cat. no.:21572-658
- Grip Size:19 mm (¾")
Spécifications
A propos de cet article
Surpass standard holding capabilities with this all-position clamp holder.
- Allows 360° rotation
- Fully adjustable holder
Building on the upgraded Q-clamp design, the holder allows adjustment at any angle in any plane. Holders are set at 90° to each other connected by a 90° connector for 360° rotation.
Customers Who Bought This Also Bought