VWR® Talon® Swivel Holder
Supplier: VWR International
Certificates
About this item
Two Q-clamp post holders with center swivel capacity allow tilting of clamp at any angle in parallel planes.
- electropolished stainless steel finish or nickel-plated zinc
- Allows tilting at any angle
- Center swivel capability
Outside adjustment screw allows close proximity between objects being held.
Customers Who Bought This Also Bought