VWR® Talon® Two- and Three-Prong Swivel Clamps
Fournisseur: VWR International
Certificats
A propos de cet article
Swivel clamps hold apparatuses near the lab frame, and unlike extension clamps, the swivel clamps have an integral holder for attaching a lab frame or other apparatus.
- electropolished stainless steel or nickel-plated zinc construction
- For circular or irregular objects
- Fully adjustable angle
- Can be locked once in place
Built-in holder grips rods up to 19mm (3/4") in diameter and is adjustable for forward or reverse-facing adjustment screws. Shaft wing nut allows the holding angle of swivel clamp to adjust through 360° of rotation and can be locked in place once desired position is acheived. Stainless steel electropolished finish or nickel-plated zinc construction.
Ordering information: Supplied with non-slip vinyl sleeves, and for temperatures above 100°C (212°F), fiberglass covers.
Customers Who Bought This Also Bought