Jazz Band® Banded Earplugs, Moldex®
# de catalogue 56610-661
Fournisseur: Moldex/Metric
Small Business Enterprise
Spécifications
- NRR:25 dB
- Description:Jazz Band® hearing protector
- Environmentally Preferable:Safer Human and Environmental Health
- Cat. no.:56610-661
- Supplier No.:6506
Spécifications
A propos de cet article
An exclusive AdB coating on the ear pods not only increases attenuation, but also repels dirt and allows the pod to be washed and reused.
- Lightweight, comfortable alternative to earmuffs
- Colorful band features soft foam ear pods
- Band will not interfere with eyewear
- Hangs easily around neck with optional cord
To use, simply position pods in ear, with band under chin (or behind head); then reach behind the head and pull top of ear upwards, push pod into ear canal with a wiggling motion to obtain a snug seal. Ear pods are easily replaced.
Customers Who Bought This Also Bought