BRAND 96-Well Microtitration Plates, BrandTech®
# de catalogue 80087-062
Fournisseur: Brandtech
Spécifications
- Sterility:Non sterile
- Description:Polypropylene Plate
- Packaging:10 Plates/Bag
- Working Volume:330 µl
- Well Shape:Round-Bottom
- Surface Treatment:Untreated
- Supplier No.:701330
Spécifications
A propos de cet article
Plates are excellent for cell and tissue culture, sample storage, immunological assays, and other applications
- ANSI/SLAS-formatted
- Polypropylene for chemical resistance
- Wells have round bottoms
Plates are SBS-formatted, alphanumerically labeled, stackable, and fully compatible with most standard plate readers and washers. Notched corners facilitate orientation in automated liquid handling systems. Raised rings around wells assist sealing with adhesive films and mats and reduce cross-contamination between wells. Nonsterile polypropylene plates are autoclavable and resistant to a wide variety of chemicals, including phenols, chloroform, and DMSO.
Customers Who Bought This Also Bought