About this item
Micro tattoo and surgery platforms can be used for surgery, micro manipulations, and general micro-utility functions such as organ inspections and dissections
Thermo-controlled surgery platform easily maintains animal body temperature during surgery with use of four space gels for continuous heat, two space blankets to retain heat, and a thermo reflector and turbo heat sink to maximize heat transfer. Thermo-controlled recovery pad enhances animal recovery with use of six space gels for continuous heat, four space blankets to retain heat, as well as a thermo reflector and turbo heat sink. Pad will not overheat or burn animal and does not require electricity or a bulky water bath.
Tail injection and tail cutting platforms feature quick restraint and are easy to clean. Tail cutting platform includes a tube holder and a ruler for accurate snips.
Both feature a non-obstructive rimless magnifying glass with a main lens magnification of 2.3x and an inset lens magnification of 4.0x. Micro tattoo platform has one flexible LED light; surgery platforms have two flexible LED lights. Both use a universal input AC power supply: 100–240V, 50/60Hz. Non-skid feet provide added stability.