A propos de cet article
For use with AIMS™ animal tattoo systems (see 89066-278 series).
Needles, made from 401 stainless steel and silver solder, are provided presterilized and ready for tattooing. Needle design minimizes animal pain and tissue damage.
Pigments have been demonstrated to be safe and nontoxic using in vivo and in vitro toxicology assays. They will not alter the physiology of animals or contaminate other lab animals in the facility.