HB-2000 Equalizer™ Earmuffs, Elvex
# de catalogue 33000-318
Fournisseur: Elvex
Small Business Enterprise
Spécifications
- NRR:25 dB
- Description:Earmuffs HB-2000 Equalizer™
- Cat. no.:33000-318
- Weight:7.4 oz.
- Supplier No.:HB-2000
Spécifications
A propos de cet article
Comfortable earmuff provides excellent noise reduction.
- Unique patented headband is designed to maintain the same tension for all head sizes
- Headband is equipped with a soft head pad for added comfort
- Soft Superseal™ ear cushions provide excellent sealing capability
- Oversized ear domes increase effectiveness against hard-to-attenuate low frequency noise
Muff contains no metal parts and is fully dielectric.
Certifications: Meets ANSI S3.19-1974 standard.
Customers Who Bought This Also Bought