ValueMuff™ Earmuffs, Elvex®
# de catalogue 33200-000
Fournisseur: Elvex
Small Business Enterprise
Spécifications
- NRR:25 dB
- Description:Earmuffs ValueMuff™
- Cat. no.:33200-000
- Supplier No.:HB-25
Spécifications
A propos de cet article
Dielectric, comfortable, lightweight muff at an economical price
- Soft, pliable Superseal™ cushions and low headband force of 2.4lbs
- Flexible slide clips for easy adjustment
- Good attenuation
Certifications: Meets ANSI 53.19-1974 standard.
Customers Who Bought This Also Bought