Leightning® Dielectric Lightweight Earmuffs, Honeywell Safety
Catalog # 55533-010
Supplier: Honeywell Safety Products
Specifications
- NRR:25 dB
- Description:Earmuffs Leightning®
- Cat. no.:55533-010
- Supplier No.:QM24+
Specifications
About this item
Dielectric three-position, ultra-lightweight earmuffs are ideal for prolonged use and comfort.
- Adjustable crown strap allows secure fit when worn behind the head or under the chin
- High-visibility red cups with black band
Weight: 170 g (6 oz.)
Customers Who Bought This Also Bought